Order Entry
Netherlands
ContactUsLinkComponent
1H-Pyrazole-3-boronic acid pinacol ester ≥98.0% (by titrimetric analysis)
Irritant
1H-Pyrazole-3-boronic acid pinacol ester ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
1H-Pyrazole-3-boronic acid pinacol ester ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    105.0 to 109.0 deg-C

Specification Test Results

Frequently Bought Together