N-([1,1':3',1''-Terphenyl]-5'-yl)-9,9-dimethyl-9H-fluoren-2-amine ≥97.0% (by HPLC, total nitrogen)
TCIAT3807-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:215.0 to 219.0 deg-C
:
:437.59 g/mol
:1372778-68-1
:C₃₃H₂₇N
:MFCD31556128