
2,4,4-Trimethylpentan-2-amine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:58618-91-0
:
Formula:C₈H₁₉N·HCl
MDL Number:MFCD30182014
:165.71 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together