
2,4,4-Trimethylpentan-2-amine hydrobromide ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1093859-61-0
:
Formula:C₈H₁₉N·HBr
:210.16 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together