N,N,N',N'-Tetrakis[4-(diisobutylamino)phenyl]-1,4-phenylenediamine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:485831-34-3
:
Formula:C₆₂H₉₂N₆
MDL Number:MFCD22207043
:921.46 g/mol
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:177.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together