

Triisocyanato(methyl)silane ≥96.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:5587-61-1
:
Formula:C₄H₃N₃O₃Si
MDL Number:MFCD00061499
:169.17 g/mol
Boiling Pt:171°C
Melting Pt:3°C
UN:2478
ADR:3,III
:GHS02
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (with Total Nitrogen):min. 96.0 %
Specification Test Results
Frequently Bought Together