4,4,5,5-Tetramethyl-2-[10-(2-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:922518-84-1
:
Formula:C₃₀H₂₇BO₂
MDL Number:MFCD28978626
:430.35 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together