
(Butylamine)[2,2'-thiobis(4-tert-octylphenolato)]nickel(II) ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14516-71-3
:
Formula:C₃₂H₅₁NNiO₂S
MDL Number:MFCD03939034
:572.52 g/mol
EINECS:238-523-3
:GHS08
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Chelometric Titration):min. 97.0 %
- Lambda max.:246.0 to 254.0 nm (CHCl3)
Specification Test Results
Frequently Bought Together