



Trimethylphenylammonium hydroxide ∼8.5% in methanol
Supplier: TCI
Certificates
About this item
CAS Number:1899-02-1
:
Formula:C₉H₁₅NO
MDL Number:MFCD00041899
ADR:3,II
:153.23 g/mol
EINECS:217-592-3
UN:2924
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid to slightly cloudy liquid
- Concentration (Neutralization titration):8.3 to 9.8 w/w%
Specification Test Results
Frequently Bought Together