Tiapride hydrochloride ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:51012-33-0
:
Formula:C₁₅H₂₄N₂O₄S·HCl
MDL Number:MFCD00133861
:364.89 g/mol
Merck Index:14,09421
Storage Temperature:Refrigerator
EINECS:256-908-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
Specification Test Results
Frequently Bought Together