
Tris(4-aminophenyl)methane ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:548-61-8
:
Formula:C₁₉H₁₉N₃
MDL Number:MFCD00137820
:289.38 g/mol
Melting Pt:208°C
EINECS:208-952-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together