5-Methylfuran-2-boronic acid pinacol ester ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:338998-93-9
:
Formula:C₁₁H₁₇BO₃
MDL Number:MFCD03094689
:208.06 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together