Order Entry
Netherlands
ContactUsLinkComponent
Tin(II) chloride ≥97.0% (by titrimetric analysis) for perovskite precursor
Corrosive
Irritant
Health
Environment
Tin(II) chloride ≥97.0% (by titrimetric analysis) for perovskite precursor
Supplier:  TCI
Certificates
Corrosive
Irritant
Health
Environment
Tin(II) chloride ≥97.0% (by titrimetric analysis) for perovskite precursor
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 97.0 %
  • Water (Vaporization methoed):
    max. 100 ppm

Specification Test Results

Frequently Bought Together