4-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:302348-51-2
:
Formula:C₁₃H₁₉BO₃
MDL Number:MFCD09837617
:234.10 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:75.0 to 79.0 deg-C
Specification Test Results
Frequently Bought Together