
Tris[2-(3-mercaptopropionyloxy)ethyl] Isocyanurate ≥70% (by HPLC)
: TCI
:36196-44-8
:
:C₁₈H₂₇N₃O₉S₃
:MFCD04038141
:525.61 g/mol
:252-907-8
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (HPLC):min. 70.0 area%
- Purity (Iodometric Titration):min. 90.0 %
Frequently Bought Together