Order Entry
Netherlands
ContactUsLinkComponent
1,3,5-Tris(4'-fluorobiphenyl-4-yl)benzene ≥95.0% (by HPLC)
1,3,5-Tris(4'-fluorobiphenyl-4-yl)benzene ≥95.0% (by HPLC)
Catalog # TCIAT3462-200MG
Supplier:  TCI
CAS Number:  
1,3,5-Tris(4'-fluorobiphenyl-4-yl)benzene ≥95.0% (by HPLC)
Catalog # TCIAT3462-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

About this item

 :
 :588.67 g/mol
Melting Pt:230°C
CAS Number:372956-40-6
Formula:C₄₂H₂₇F₃