N,N,N',N'-Tetrakis(4-nitrophenyl)-1,4-phenylenediamine ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:3283-05-4
:
Formula:C₃₀H₂₀N₆O₈
MDL Number:MFCD23135517
:592.52 g/mol
Melting Pt:392°C
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together