5-(p-Tolyl)isoxazole-3-carboxaldehyde ≥98.0% (by GC)
Catalog # TCIAT3454-1G
Leverancier: TCI
CAS-nummer:
Specificaties
- Pk:1 g
- Seq:3047
Specificaties
Specificatie testresultaten
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:70.0 to 74.0 deg-C
- Acid Value:max. 2.0
Specificatie testresultaten
Over dit artikel
:
:187.20 g/mol
CAS-nummer:640292-02-0
Formule:C₁₁H₉NO₂
MDL-nummer:MFCD06199344