
α,α,α′,α′-Tetramethyl-1,3-benzenedipropionic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAT3420-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:278.35 g/mol
Melting Pt:155°C
:GHS07
CAS Number:819050-88-9
Formula:C₁₆H₂₂O₄
MDL Number:MFCD08276874