4-(p-Tolyl)cyclohexanone ≥98.0% (by GC)
Catalog # TCIAT3418-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:49.0 to 53.0 deg-C
Specification Test Results
About this item
:
:188.27 g/mol
CAS Number:40503-90-0
Formula:C₁₃H₁₆O
MDL Number:MFCD14584461