
1-(2-Tetrahydrofurfuryl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
TCIAT3413-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:95.0 to 99.0 deg-C
:
:160.24 g/mol
:2811
:6.1,III
:GHS06
:66892-25-9
:C₆H₁₂N₂OS
:MFCD00060498