
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1009033-87-7
:
Formula:C₁₇H₂₀BNO₂
MDL Number:MFCD12828171
:281.16 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:110.0 to 114.0 deg-C
Specification Test Results
Frequently Bought Together