Order Entry
Netherlands
ContactUsLinkComponent
Trimethyl[[(1E,3E)-2-methylpenta-1,3-dien-1-yl]oxy]silane ≥95.0% (by GC)
Flammable
Irritant
Trimethyl[[(1E,3E)-2-methylpenta-1,3-dien-1-yl]oxy]silane ≥95.0% (by GC)
Catalog # TCIAT3396-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Trimethyl[[(1E,3E)-2-methylpenta-1,3-dien-1-yl]oxy]silane ≥95.0% (by GC)
Catalog # TCIAT3396-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :170.33 g/mol
Boiling Pt:68°C
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:82223-91-4
Formula:C₉H₁₈OSi