
3,4,5-Trichlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:862248-93-9
:
Formula:C₆H₄BCl₃O₂
MDL Number:MFCD03095126
:225.27 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 109.0 %
Specification Test Results
Frequently Bought Together