
2,4,6-Trichlorophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT3382-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 108.0 %
Specification Test Results
About this item
:
:225.26 g/mol
:GHS07
CAS Number:73852-18-3
Formula:C₆H₄BCl₃O₂
MDL Number:MFCD01074621