Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
[3-(Trimethoxysilyl)propyl]succinic anhydride ≥95.0% (by titrimetric analysis)
Catalog # TCIAT3360-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
[3-(Trimethoxysilyl)propyl]succinic anhydride ≥95.0% (by titrimetric analysis)
Catalog # TCIAT3360-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 93.0 %
  • Purity (Morpholine Method):
    min. 95.0 %

Specification Test Results

About this item

 :
 :262.33 g/mol
Flash Pt:190°C
 :GHS07
CAS Number:156088-53-8
Formula:C₁₀H₁₈O₆Si