
3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)
: TCI
:939430-30-5
:
:C₁₇H₂₀BNO₂
:MFCD20923943
:281.16 g/mol
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:95.0 to 99.0 deg-C
Frequently Bought Together