
2-Naphthyl boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:256652-04-7
:
Formula:C₁₆H₁₉BO₂
MDL Number:MFCD05663868
:254.14 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:64.0 to 68.0 deg-C
Specification Test Results
Frequently Bought Together