N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:122738-21-0
:
Formula:C₄₀H₃₆N₂O₄
MDL Number:MFCD09833415
:608.74 g/mol
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:190.0 to 194.0 deg-C
Specification Test Results
Frequently Bought Together