
4,4,5,5-Tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane ≥95.0% (by HPLC, titration analysis)
: TCI
:1143576-84-4
:
:C₃₂H₂₉BO₂
:456.39 g/mol
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
Frequently Bought Together