
4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1115639-92-3
:
Formula:C₃₀H₂₇BO₂
MDL Number:MFCD28130383
:430.35 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:177.0 to 181.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together