
Tetrabutylammonium bis(4-methyl-1,2-benzenedithiolato)nickelate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15492-42-9
:
Formula:C₃₀H₄₈NNiS₄
MDL Number:MFCD12022530
:609.65 g/mol
:GHS08
Specification Test Results
- Appearance:Dark green to Dark red to Black powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Chelometric Titration):min. 98.0 %
- Melting point:145.0 to 150.0 deg-C
Specification Test Results
Frequently Bought Together