Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
(3-Acetamidophenyl)boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAT3216-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(3-Acetamidophenyl)boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAT3216-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    188.0 to 192.0 deg-C

Specification Test Results

About this item

 :
 :261.1245 g/mol
 :GHS07
CAS Number:480424-93-9
Formula:C₁₄H₂₀BNO₃
MDL Number:MFCD03789261