
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:903550-26-5
:
Formula:C₁₄H₂₃BN₂O₃
MDL Number:MFCD09037501
:278.16 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:74.0 to 78.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together