
2,4,5-Trifluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:220227-21-4
:
Formula:C₆H₂ClF₃O₂S
MDL Number:MFCD00077600
:230.59 g/mol
Flash Pt:100°C
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together