Order Entry
Netherlands
ContactUsLinkComponent
[[(3,4,5-Trimethyl-1H-pyrrol-2-yl)(3,4,5-trimethyl-2H-pyrrol-2-ylidene)methyl]carbonitrile](difluoroborane) ≥98.0% (by GC)
Irritant
[[(3,4,5-Trimethyl-1H-pyrrol-2-yl)(3,4,5-trimethyl-2H-pyrrol-2-ylidene)methyl]carbonitrile](difluoroborane) ≥98.0% (by GC)
Catalog # TCIAT3127-200MG
Supplier:  TCI
CAS Number:  
Irritant
[[(3,4,5-Trimethyl-1H-pyrrol-2-yl)(3,4,5-trimethyl-2H-pyrrol-2-ylidene)methyl]carbonitrile](difluoroborane) ≥98.0% (by GC)
Catalog # TCIAT3127-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :301.15 g/mol
 :GHS07
CAS Number:157410-23-6
Formula:C₁₆H₁₈BF₂N₃