Order Entry
Netherlands
ContactUsLinkComponent
1-(p-Tolyl)-1-cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(p-Tolyl)-1-cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
1-(p-Tolyl)-1-cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    169.0 to 173.0 deg-C

Specification Test Results

Frequently Bought Together