Order Entry
Netherlands
ContactUsLinkComponent
 
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate ≥98.0% (by GC)
 :  TCI
 
undefined
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Yellow to Orange powder to lump
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together