
Tris(2-methoxyphenyl)phosphine ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:4731-65-1
:
Formula:C₂₁H₂₁O₃P
MDL Number:MFCD00014892
:352.37 g/mol
EINECS:225-235-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Iodometric Titration):min. 95.0 %
- Melting point:202.0 to 206.0 deg-C
Specification Test Results
Frequently Bought Together