L-Tryptophan ethyl ester hydrochloride ≥95.0% (by HPLC, titration analysis)
Catalogus # TCIAT2981-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Specific rotation [a]20/D:+10.0 to +13.0 deg (C=1, MeOH)
Specification Test Results
About this item
:
:268.74 g/mol
EINECS:220-786-0
CAS Number:2899-28-7
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD00038993