β-Thionicotinamide adenine dinucleotide, reduced form disodium salt ≥93.0% (by HPLC) for biochemical research
Catalog # TCIAT2980-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 93.0 area%
- Enzyme detection test:to pass test
Specification Test Results
About this item
:
:725.47 g/mol
Storage Temperature:Freezer
CAS Number:1921-48-8
Formula:C₂₁H₂₇N₇O₁₃P₂S·2Na
MDL Number:MFCD00133854