O-TBDPS-D-Thr-N-Boc-L-tert-Leu-Diphenylphosphine ≥98.0% (by HPLC)
TCIAT2937-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):3.60 to 4.00 %
- Specific rotation [a]20/D:-26.0 to -29.0 deg (C=1、CHCl3)
:
:725.00 g/mol
:1264520-63-9
:C₄₃H₅₇N₂O₄PSi