
Triphenylmethylium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:136040-19-2
:
Formula:C₄₃H₁₅BF₂₀
MDL Number:MFCD03426981
:922.37 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together