BAPTA-AM ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:126150-97-8
:
Formula:C₃₄H₄₀N₂O₁₈
MDL Number:MFCD00036696
:764.69 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:96.0 to 100.0 deg-C
Specification Test Results
Frequently Bought Together