Order Entry
Netherlands
ContactUsLinkComponent
3,4,5-Trimethoxyphenylboronic acid (contains varying amounts of Anhydride)
Irritant
3,4,5-Trimethoxyphenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2837-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3,4,5-Trimethoxyphenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2837-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Neutralization titration):
    97.0 to 110.0 %

Specification Test Results

About this item

 :
 :212.01 g/mol
Melting Pt:247°C
 :GHS07
CAS Number:182163-96-8
Formula:C₉H₁₃BO₅
MDL Number:MFCD01075676