
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl alcohol ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:87736-88-7
:
Formula:C₉H₇F₃N₂O
MDL Number:MFCD19442582
:216.16 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to lump
- Purity (HPLC):min. 97.0 area%
Specification Test Results
Frequently Bought Together