
2-(Trifluoromethyl)phenylhydrazinium chloride ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:3107-34-4
:
Formula:C₇H₈ClF₃N₂
MDL Number:MFCD00102619
:212.6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Potassium iodate Method):min. 97.0 %
Specification Test Results
Frequently Bought Together