Order Entry
Netherlands
ContactUsLinkComponent
2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene ≥98.0% (by HPLC)
Irritant
2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene ≥98.0% (by HPLC)
Catalog # TCIAT2759-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene ≥98.0% (by HPLC)
Catalog # TCIAT2759-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal

Specification Test Results

About this item

 :
 :644.00 g/mol
Melting Pt:455°C
 :GHS07
CAS Number:27192-91-2
Formula:C₂₆H₁₂Br₄
MDL Number:MFCD22581445