

1H-Tetrazole-5-acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:21743-75-9
:
Formula:C₃H₄N₄O₂
MDL Number:MFCD03094010
:128.09 g/mol
Melting Pt:177°C
EINECS:244-563-2
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together