Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAT2706-5G
Supplier:  TCI
CAS Number:  
Irritant
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAT2706-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomer)

Specification Test Results

About this item

 :
 :168.16 g/mol
Boiling Pt:181°C
 :GHS07
CAS Number:30129-18-1
Formula:C₇H₁₁F₃O
MDL Number:MFCD00102144